

Zirconyl chloride hydrate ≥99.9% (metals basis)
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:15461-27-5
:
Formula:Cl₂OZr·H₂O
MDL Number:MFCD00149897
:196.14 g/mol
Merck Index:14,10184
Melting Pt:400 °C (dec.)
Density:1.91
Storage Temperature:Ambient
EINECS:231-717-9
UN:3260
ADR:8,II
:GHS05
:GHS07