(S)-(-)-2,2'-Bis(diphenylphosphino)-1,1'-binaphthyl
Catalog # MOLE28868113-1G
Supplier: Molekula
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:622.69 g/mol
Merck Index:13,01223
Melting Pt:241…242 °C
Storage Temperature:Ambient
CAS Number:76189-56-5
Formula:C₄₄H₃₂P₂
MDL Number:MFCD00010805