Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
kr 0,00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Norway
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Gallic acid monohydrate
Gallic acid monohydrate
Gallic acid monohydrate
MOLE19107161-1KG
:
Molekula
:
Gallic acid monohydrate
MOLE19107161-1KG
:
Molekula
:
Pack type:
Plastic bottle for solids
Pk:
1 kg
Seq:
4024
:
:
188.14 g/mol
:
∼250 °C
:
250 °C (closed cup)
:
205-749-9
:
GHS07
:
Ambient
:
5995-86-8
:
C₇H₆O₅·1H₂O
:
MFCD00149098