Order Entry
Norway
ContactUsLinkComponent
 
 
 
Sodium hydroxide 6 N in aqueous solution, BAKER ANALYZED® volumetric solution, J.T.Baker®
  5672-02
 :  Avantor
 :  
undefined
 
 
Sodium hydroxide 6 N in aqueous solution, BAKER ANALYZED® volumetric solution, J.T.Baker®
  5672-02
 :  Avantor
 :  

 

  • Pack type:
    PE bottle, narrow mouth
  • Pk:
    1 L
  • Seq:
    5037

 

 

  • For Laboratory,Research,or Manufacturing Use:
    5.98 - 6.02
  • Protect from air to avoid absorption of carbon dioxide.:
    Max. 5 ppm
  • Normality:
    5.98 - 6.02
  • Chloride (Cl):
    ≤5 ppm
  • ACS - Heavy Metals (as Pb):
    ≤1 ppm
  • Trace Impurities - Iron (Fe):
    ≤0.5 ppm

 

 

 :
 :40 g/mol
 :109 °C (1013 hPa)
 :≤1.11 g/cm³ (20 °C)
 :Ambient
 :215-185-5
 :1824
 :8,II
 :GHS05
 :GHS07
 :1310-73-2
 :NaOH
 :MFCD00003548