Order Entry
Norway
ContactUsLinkComponent
Potassium iodate-iodide (0.713 g/L KIO₃) 0.02 N in aqueous solution stabilised volumetric solution
Potassium iodate-iodide (0.713 g/L KIO₃) 0.02 N in aqueous solution stabilised volumetric solution
Supplier:  VWR Chemicals
Certificates
undefined
Potassium iodate-iodide (0.713 g/L KIO₃) 0.02 N in aqueous solution stabilised volumetric solution
Supplier:  VWR Chemicals
Certificates
Specification Test Results
  • Assay (Potassium iodate):
    0.7126 - 0.7140 g/l