Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
kr 0,00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Norway
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Phenol liquified
Phenol liquified ≥80%
Phenol liquified ≥80%
294785A
:
VWR Chemicals
:
Phenol liquified ≥80%
294785A
:
VWR Chemicals
:
Pack type:
Glass bottle, Safebreak®
Pk:
2,5 L
Seq:
5096
Assay:
78.5 - 81.5 %
:
:
94.11 g/mol
:
Ambient
:
203-632-7
:
2821
:
6.1,II
:
GHS06
:
GHS05
:
GHS08
:
108-95-2
:
C₆H₅OH
:
MFCD00002143