Order Entry
Norway
ContactUsLinkComponent
Ferroin (1,10-phenanthrolineferrous sulphate complex) (7 mg/ml FeSO₄) in aqueous solution, AVS TITRINORM volumetric solution, for COD determination according to NF T 90-101 standard
Ferroin (1,10-phenanthrolineferrous sulphate complex) (7 mg/ml FeSO₄) in aqueous solution, AVS TITRINORM volumetric solution, for COD determination according to NF T 90-101 standard
Catalog # 30890.188
Supplier:  VWR Chemicals
CAS Number:  
undefined
Ferroin (1,10-phenanthrolineferrous sulphate complex) (7 mg/ml FeSO₄) in aqueous solution, AVS TITRINORM volumetric solution, for COD determination according to NF T 90-101 standard
Catalog # 30890.188
Supplier:  VWR Chemicals
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    100 mL
  • Seq:
    2440

Specifications

Specification Test Results

  • Identification:
    Passes test

Specification Test Results

About this item

 :
 :692.54 g/mol
EINECS:238-676-6
CAS Number:14634-91-4
Formula:C₃₆H₂₄FeN₆SO₄
MDL Number:MFCD00036428