Order Entry
Norway
ContactUsLinkComponent
Fibrowax (formulation Raymond A. Lamb), in pastille form
Irritant
Fibrowax (formulation Raymond A. Lamb), in pastille form
Catalog # 361427G
Supplier:  VWR Chemicals
CAS Number:  
undefined
Irritant
Fibrowax (formulation Raymond A. Lamb), in pastille form
Catalog # 361427G
Supplier:  VWR Chemicals
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    10 kg
  • Seq:
    4158

Specifications

Specification Test Results

  • Appearance (65°C):
    Passes test
  • Cutting/ribboning:
    Passes test
  • Congealing range:
    55 - 58 °C

Specification Test Results

About this item

 :
 :GHS07
Storage Temperature:Ambient