Order Entry
Norway
ContactUsLinkComponent
 
Paraffin wax, Solidification point: 55 - 58 °C, in pastille form, Gurr™
  361077E
 :  
undefined
Paraffin wax, Solidification point: 55 - 58 °C, in pastille form, Gurr™
  361077E
 :  

 

  • Pack type:
    Plastic bag
  • Pk:
    10 kg
  • Seq:
    4158

 

 

  • Appearance (65 °C):
    Passes test
  • Congealing range:
    55 - 58 °C
  • Cutting/ ribboning:
    Passes test

 

 

 :
 :0.9 g/cm³ (20 °C)
 :>100 °C
 :Ambient
 :8002-74-2
 :64742-51-4