Order Entry
Norway
ContactUsLinkComponent
Anti-IgG Goat Antibody (CyDye™ 800)
Anti-IgG Goat Antibody (CyDye™ 800)
Supplier:  Cytiva
Certificates
undefined
Anti-IgG Goat Antibody (CyDye™ 800)
Supplier:  Cytiva

Specifications

  • Catalog No:
  • 29-3607-89
  • 29-3607-88
  • Antigen symbol:
  • IgG
  • IgG
  • Antibody type:
  • Secondary
  • Secondary
  • Conjugation:
  • CyDye™ 800
  • CyDye™ 800
  • Reactivity:
  • Mouse
  • Mouse
  • Host:
  • Goat
  • Goat
  • Western blot:
  • Yes
  • Yes
  • ImmunoFluorescence:
  • Yes
  • Yes
  • Storage temperature:
  • 2...8 °C
  • 2...8 °C

Specifications

Related Information