Order Entry
Norway
ContactUsLinkComponent
Ethyl Gallate, for Determination of Total Polyphenol Content
Ethyl Gallate, for Determination of Total Polyphenol Content
Catalog # TCIAE0854-5G
Supplier:  TCI
CAS Number:  
undefined
Ethyl Gallate, for Determination of Total Polyphenol Content
Catalog # TCIAE0854-5G
Supplier:  TCI
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    5 g
  • CAS No.:
    831-61-8
  • Description:
    Ethyl Gallate, Purity (Neutralization titration): min. 98.0 %
  • Melting point:
    151,0…155,0 °C
  • Formula:
    C₉H₁₀O₅
  • Description:
    Ethyl Gallate

Specifications

About this item