Order Entry
Norway
ContactUsLinkComponent
Acrylonitrile ≥99.0% stabilised, GPR RECTAPUR®
Acrylonitrile ≥99.0% stabilised, GPR RECTAPUR®
  87092.290
 :  
undefined
Acrylonitrile ≥99.0% stabilised, GPR RECTAPUR®
  87092.290
 :  

 

  • Pack type:
    Glass bottle
  • Pk:
    1 L
  • Seq:
    5037

 

 

  • Assay :
    Min. 99,0 %
  • Stabilizer MEHQ 35-45 ppm:
    Confirmed
  • Density (20/4):
    0,805 - 0,807
  • Refractive index (20° C; 589 nm):
    1,390 - 1,392
  • Water:
    ≤ 1.0 %

 

 

 :
 :53.06 g/mol
 :–5 °C
 :1093
 :3,I
 :Stabilised with hydroquinone monomethyl ether (4-methoxyphenol, MEHQ) 35-45 ppm
 :203-466-5
 :GHS02
 :GHS06
 :GHS05
 :GHS08
 :GHS09
 :107-13-1
 :C₃H₃N
 :MFCD00001927