General purpose gloves, Showa 381
: Showa
SHOWA 381 is a general purpose, durable, lightweight, foam nitrile coated, with a breathable microfibre knit liner and extremely comfortable gloves. Ideal for assembly and precision handling tasks.
- Microporous nitrile coating allows warm air and moisture from inside to escape
- Excellent grip in light oily and dry environments
- Microfibre yarn absorbs perspiration and moisture from the skin, resulting in drier and cooler hands
- Excellent levels of tactility and sensitivity
- Exceptional suppleness, comfort and fit
- Abrasion level 4 for maximum abrasion resistance
Ideal for use in construction, maintenance, logistics, aerospace, automotive, engineering and manufacturing. Typical applications include: Assembly of dry and oily parts, general handling, handling of intricate components, light fabrication, mounting and dismounting of parts and tool setting.
: EN 388:2016 (4121X); Cat. II
- Catalog No:
- 112-5827
- 112-5826
- 112-5829
- 112-5828
- 112-5825
- Model:
- Showa 381
- Showa 381
- Showa 381
- Showa 381
- Showa 381
- Ambidextrous / hand-specific:
- Hand-specific
- Hand-specific
- Hand-specific
- Hand-specific
- Hand-specific
- Disposable / reusable:
- Reusable
- Reusable
- Reusable
- Reusable
- Reusable
- Latex / latex-free:
- Latex-free
- Latex-free
- Latex-free
- Latex-free
- Latex-free
- Finish:
- Embossed microporous nitrile coating (palm)
- Embossed microporous nitrile coating (palm)
- Embossed microporous nitrile coating (palm)
- Embossed microporous nitrile coating (palm)
- Embossed microporous nitrile coating (palm)
- Gauge:
- 13
- 13
- 13
- 13
- 13
- Cuff style:
- Knit wrist
- Knit wrist
- Knit wrist
- Knit wrist
- Knit wrist
- EN norm:
- EN 388:2016 (4121X)
- EN 388:2016 (4121X)
- EN 388:2016 (4121X)
- EN 388:2016 (4121X)
- EN 388:2016 (4121X)