General purpose gloves, Showa 306
Supplier: Showa
Certificates
About this item
These outdoor gloves are impermeable yet breathable to keep hands dry whilst preventing loss of grip from perspiration.
- Durable fully-coated latex
- Heavy duty handling tasks
- Can be exposed to changeable weather conditions
- Reduced perspiration when it's hot yet warmth and dryness in wet conditions
Seamless 13 gauge Nylon/polyester knitted liner provides powerful grip, with breathable coating technology and long lasting grip performance in wet and dry conditions. The anatomical design helps prevent hand fatigue and aerated foam latex provides cushioning from minor impacts and with soft and comfortable fit.
SHOWA 306 series gloves are ideally suited for aerospace, agriculture, automotive, construction, highway maintenance, manufacturing, railways and wet work.
Certifications: EN 388:2016 (2121X); Cat. II
Packaging: 10 pairs per pack.
Specifications
- Catalog No:
- 112-5760
- 112-5764
- 112-5765
- 112-5766
- 112-5759
- Model:
- Showa 306
- Showa 306
- Showa 306
- Showa 306
- Showa 306
- Ambidextrous / hand-specific:
- Hand-specific
- Hand-specific
- Hand-specific
- Hand-specific
- Hand-specific
- Latex / latex-free:
- Latex
- Latex
- Latex
- Latex
- Latex
- Liner material:
- Nylon/PE
- Nylon/PE
- Nylon/PE
- Nylon/PE
- Nylon/PE
- Finish:
- Palm and fingertip coated
- Palm and fingertip coated
- Palm and fingertip coated
- Palm and fingertip coated
- Palm and fingertip coated
- Gauge:
- 13
- 13
- 13
- 13
- 13
- Cuff style:
- Knitted elasticated wrist and cuff
- Knitted elasticated wrist and cuff
- Knitted elasticated wrist and cuff
- Knitted elasticated wrist and cuff
- Knitted elasticated wrist and cuff
- EN norm:
- EN 388:2016 (2121X)
- EN 388:2016 (2121X)
- EN 388:2016 (2121X)
- EN 388:2016 (2121X)
- EN 388:2016 (2121X)