Hand disinfectants, Swarfega® Orange
This is a solvent-free workshop cream with plastic microbeads, effective against strong and adherent soiling such as greases, oils and fuels.
- Glycerin and wheat germ oil increase skin tolerance and hydrate the skin
- Fresh fragrance based on natural orange extracts
- Solvent-free form helps to prevent skin dryness
- Contains glycerin and wheat germ extracts to help hydration of cutaneous skin
Contains micropolymer granules which provide deep cleaning and gently loosen crusted contaminants without damaging the skin. This rich and creamy lotion rinses off quickly, without leaving an oily feeling on the skin. A subtle orange scent leaves the skin fresh and slightly scented after use.
Applications include heavy industry, general and light industry.
: *The product(s) on this page are outside the scope of the EU Directive 2019/904