J.T.Baker®, Proclean
Catalog # 3768.1000
Supplier: Avantor
CAS Number:
Specifications
- Pk:1 L
- Description:Proclean, micros bottle
Specifications
About this item
pH at 25 °C: 8,7 to 8,9
Total Bacterial Growth (CFU/100 ml): Max. 5
pH at 25 °C: 8,7 to 8,9
Total Bacterial Growth (CFU/100 ml): Max. 5