Naloxone N-oxide solution, certified reference material, Supelco®
Catalog # IMPN-007-04-1M
Supplier: Merck
CAS Number:
Specifications
- Concentration:100 mug/mL in acetonitrile: water (1:1)
- Pk:1 mL
- Standards:certified reference material
- CAS No.:112242-14-5
- Description:Naloxone N-oxide solution
- Description:Naloxone N-oxide solution
- Pk Info:ampoule of 1 ml