Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
kr 0,00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Norway
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Standards
Organic Standards
4-Hydroxyestradiol, analytical standard, Supelco®
4-Hydroxyestradiol, analytical standard, Supelco®
4-Hydroxyestradiol, analytical standard, Supelco®
Catalog # 41171-2MG
Supplier:
Merck
CAS Number:
Expand image
4-Hydroxyestradiol, analytical standard, Supelco®
Catalog # 41171-2MG
Supplier:
Merck
CAS Number:
Specifications
Pk:
2 mg
Standards:
analytical standard
CAS No.:
5976-61-4
Description:
4-Hydroxyestradiol
Description:
4-Hydroxyestradiol
Specifications
About this item
Product Details
Product Details