Order Entry
Norway
ContactUsLinkComponent
Supelco® Organic Reference Standard, Trans-Isoferulic Acid
Supelco® Organic Reference Standard, Trans-Isoferulic Acid
Catalog # 05407-10MG
Supplier:  Merck
CAS Number:  
undefined
Supelco® Organic Reference Standard, Trans-Isoferulic Acid
Catalog # 05407-10MG
Supplier:  Merck
CAS Number:  

Specifications

  • Pk:
    10 mg
  • Standards:
    Analytical standard
  • CAS No.:
    25522-33-2
  • Description:
    trans-Isoferulic acid
  • Storage conditions:
    2...8 °C
  • Hazard class description:
    H315 - H319 - H335
  • Shipping conditions:
    STD
  • MDL No.:
    MFCD00004391
  • M-Clarity quality level:
    MQ100
  • Synonym(s):
    trans-3-(3-Hydroxy-4-methoxyphenyl)acrylic acid; trans-3-Hydroxy-4-methoxycinnamic acid; Hesperetic acid
  • Melting point:
    230...236 °C
  • Formula:
    C₁₀H₁₀O₄
  • Molecular weight:
    194,18
  • Storage temperature:
    2...8 °C
  • Flash point:
    Not applicable

Specifications

About this item

Packaging: Bottomless glass bottle. Contents are inside inserted fused cone.
Caution: P261 - P264 - P271 - P280 - P302 + P352 - P305 + P351 + P338