Order Entry
Norway
ContactUsLinkComponent
Supelco® Organic Reference Standard, Neryl acetate
Supelco® Organic Reference Standard, Neryl acetate
Catalog # 46015-1ML
Supplier:  Merck
CAS Number:  
undefined
Supelco® Organic Reference Standard, Neryl acetate
Catalog # 46015-1ML
Supplier:  Merck
CAS Number:  

Specifications

  • Pk:
    1 mL
  • Standards:
    Analytical standard
  • CAS No.:
    141-12-8
  • Description:
    Neryl acetate
  • Storage conditions:
    2…8 °C
  • Shipping conditions:
    STD
  • MDL No.:
    MFCD00063205
  • M-Clarity quality level:
    MQ100
  • Synonym(s):
    cis-3,7-Dimethyl-2,6-octadien-1-yl acetate
  • Formula:
    CH₃CO₂CH₂CH=C(CH₃)CH₂CH₂CH=C(CH₃)₂
  • Molecular weight:
    196,29
  • Density:
    0,91 g/ml at 25 °C (lit.)
  • Flash point:
    99 °C - Closed cup
  • Boiling point:
    134 °C/25 mmHg (lit.)

Specifications

About this item