Supelco® Organic Reference Standard, Bradykinin acetate salt
Catalog # 90834-10MG
Supplier: Merck
CAS Number:
Specifications
- Pk:10 mg
- Standards:Analytical standard
- CAS No.:6846-03-3
- Description:Bradykinin acetate salt
- Storage conditions:2…8 °C
- Shipping conditions:STD
- M-Clarity quality level:MQ100
- Molecular weight:1060,21 (free base basis)