Order Entry
Norway
ContactUsLinkComponent
Hexyl chloroformate ≥97.0% (by GC), LiChropur™ for GC derivatisation, Supelco®
Hexyl chloroformate ≥97.0% (by GC), LiChropur™ for GC derivatisation, Supelco®
Catalog # 67389-10ML
Supplier:  Merck
CAS Number:  
undefined
Hexyl chloroformate ≥97.0% (by GC), LiChropur™ for GC derivatisation, Supelco®
Catalog # 67389-10ML
Supplier:  Merck
CAS Number:  

Specifications

  • Pk:
    10 mL
  • Seq:
    2275
  • Pk Info:
    1× 10 ml

Specifications

About this item

 :
 :164.6299 g/mol
CAS Number:6092-54-2
Formula:C₇H₁₃ClO₂
MDL Number:MFCD00000651