Organic reference standard, Triphenylphosphine oxide, analytical standard, Supelco®
08902-100MG
: Merck
:
- Pk:100 mg
- Standards:Analytical standard
- CAS No.:791-28-6
- Description:Triphenylphosphine oxide
- Melting point:150...157 °C (lit.)
- Formula:(C₆H₅)₃PO
- Description:Triphenylphosphine oxide