Supelco® PESTANAL® Organic Reference Standard, Ethyl acetate
45758-100MG-R
: Merck
:
- Fp:100 mg
- Standard:PESTANAL®, analytical standard
- CAS-nr:141-78-6
- Beskrivelse:Ethyl acetate
- Lagringsforhold:2...30 °C
- Hardhetsklasse beskrivelse:H412
- Fraktforhold:STD
- MDL No.:MFCD00009171
- M-Clarity quality level:MQ100
- Synonym(s):EtOAc
- Formel:CH₃COOC₂H₅
- Molekylvekt:88,11
: P273 - P501