Order Entry
Norway
ContactUsLinkComponent
Organic reference standard, clofibric acid, analytical standard, Supelco®
Organic reference standard, clofibric acid, analytical standard, Supelco®
Catalog # 90323-100MG
Supplier:  Merck
CAS Number:  
undefined
Organic reference standard, clofibric acid, analytical standard, Supelco®
Catalog # 90323-100MG
Supplier:  Merck
CAS Number:  

Specifications

  • Pk:
    100 mg
  • Standards:
    Analytical standard
  • CAS No.:
    882-09-7
  • Description:
    Clofibric acid
  • Melting point:
    120...122 °C (lit.)
  • Formula:
    C₁₀H₁₁ClO₃
  • Description:
    Clofibric acid

Specifications

About this item