Order Entry
Norway
ContactUsLinkComponent
Supelco® Organic Reference Standard, cis-8,11,14-Eicosatrienoic acid
Supelco® Organic Reference Standard, cis-8,11,14-Eicosatrienoic acid
Catalog # 69154-50MG
Supplier:  Merck
CAS Number:  
undefined
Supelco® Organic Reference Standard, cis-8,11,14-Eicosatrienoic acid
Catalog # 69154-50MG
Supplier:  Merck
CAS Number:  

Specifications

  • Pk:
    50 mg
  • Standards:
    Analytical standard
  • CAS No.:
    1783-84-2
  • Description:
    cis-8,11,14-Eicosatrienoic acid
  • Storage conditions:
    –20 °C
  • Shipping conditions:
    WICE
  • MDL No.:
    MFCD00065721
  • M-Clarity quality level:
    MQ100
  • Synonym(s):
    Homo-γ-linolenic acid
  • Formula:
    CH₃(CH₂)₃(CH₂CH=CH)₃(CH₂)₆CO₂H
  • Molecular weight:
    306,48
  • Flash point:
    62 °C - Closed cup

Specifications

About this item