Order Entry
Norway
ContactUsLinkComponent
Pesticide reference standard, 2,3-Dichlorophenoxyacetic acid, PESTANAL®, analytical standard, Supelco®
Pesticide reference standard, 2,3-Dichlorophenoxyacetic acid, PESTANAL®, analytical standard, Supelco®
Catalog # 92397-50MG
Supplier:  Merck
CAS Number:  
undefined
Pesticide reference standard, 2,3-Dichlorophenoxyacetic acid, PESTANAL®, analytical standard, Supelco®
Catalog # 92397-50MG
Supplier:  Merck
CAS Number:  

Specifications

  • Pk:
    50 mg
  • Standards:
    Analytical standard
  • CAS No.:
    2976-74-1
  • Description:
    2,3-Dichlorophenoxyacetic acid
  • Melting point:
    170...175 °C
  • Formula:
    C₈H₆Cl₂O₃
  • Description:
    2,3-Dichlorophenoxyacetic acid

Specifications

About this item