Kalsiumhydrogenfosfat, white powder
Supplier: MP Biomedicals
Certificates
About this item
CAS Number:7757-93-9
:
Formula:CaHPO₄
MDL Number:MFCD00010909
:136.06 g/mol
Melting Pt:∼370 °C (dec., lit.)
Density:∼2,3 g/cm³ (20 °C) (lit.)
Storage Temperature:Romtemperatur
EINECS:231-826-1
Specification Test Results
- Identity Test :Passes