Test strips, qualitative, iron
Iron Test paper for Iron, 10 - mg/L - Fe²⁺ or Fe³⁺, Complexation with a catechol, 48×25×84 mm (package), 100 test(s), 100
Supplier: MACHEREY-NAGEL
Jump to:
Product Details & Documents
This test paper is used for the rapid detection of Iron ions (Fe₂⁺ and Fe⁺). In contrast to Dipyridylpaper it reacts with Fe(II) as well as Fe(III) ions. For the specific detection of Fe(II) please use Dipyridylpaper.
- Colour change: Yellow-white to red-brown
Limit of sensitivity: 10 mg/L Fe₂⁺ or Fe₃⁺
Packaging: Pack of 100 strips 20×70 mm
Documents
Specifications
Specifications
- Chemical method:Complexation with a catechol
- Pk:100
- Description:Iron Test paper for Iron
- Dimensions:48×25×84 mm (package)
- Tests per kit:100 test(s)
- Range:10 - mg/L - Fe²⁺ or Fe³⁺
- Cat. No.:MANA90726
- Sensitivity:Iron - 10 mg/L Fe²⁺ or Fe³⁺
- Contents:100 test strips in a plastic box
- Platform:Qualitative test paper
- Compliance:Not CE certified and no hazardous material
- Shelf life (months):24 (from production)
- W×D×H:20×70 mm (net)
- Method:Iron - yellow-white → red-brown
- Weight:44,5 g



