Order Entry
Norway
Orders LinkContactUsLinkComponent
Arsenic Qualitative Test Papers

Arsenic Qualitative Test Papers

Antimony test paper for antimony, 5 - mg/L - Sb³⁺, Complexation with a fluorone, 48×25×84 mm (packaging), 200 Tests, 200
Catalog # MANA90723
Supplier:   MACHEREY-NAGEL
Arsenic Qualitative Test Papers
Catalog # MANA90723
Supplier:  MACHEREY-NAGEL
CAS Number:  
Arsenic Qualitative Test Papers
Arsenic Qualitative Test Papers

Antimony test paper for antimony, 5 - mg/L - Sb³⁺, Complexation with a fluorone, 48×25×84 mm (packaging), 200 Tests, 200

CAS Number:  
Price
kr 781,00
Log in for å se tilgjengelighet og deres pris
1 * 200 Items
Product Details & Documents

This test paper allows the easy detection of arsine (AsH₃) in the gas phase. Arsenic in solutions has to be converted into AsH₃ with Zn/acid and purged from the solution. The Arsine is detected directly at the boundary layer between water and air.

  • Colour change: White to brown-black

Arsenic test paper is used for the easy determination of arsenic in grape must and wine. For the determination of Arsine in compartment air the test paper is moistened with acetic anhydride.

Spesifikasjoner

Spesifikasjoner

  • Chemical method:
    Complexation with a fluorone
  • Pk:
    200
  • Description:
    Antimony test paper for antimony
  • Dimensions:
    48×25×84 mm (packaging)
  • Tests per kit:
    200 Tests
  • Range:
    5 - mg/L - Sb³⁺
  • Cat. No.:
    MANA90723
  • Sensitivity:
    Antimony - 5 mg/L Sb³⁺
  • Contents:
    200 test strips in a plastic box
  • Platform:
    Qualitative test paper
  • Compliance:
    Not CE certified and no hazardous material
  • Shelf life (months):
    24 (from production)
  • W×D×H:
    20×70 mm (net)
  • Method:
    Antimony - yellow → orange-red
  • Weight:
    48,24 g

Spesifikasjoner

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.