Order Entry
Norway
ContactUsLinkComponent
Molecular sieve dehydrate, beads for drying gases, with indicator, Ø 2-5 mm, Fluka™
Molecular sieve dehydrate, beads for drying gases, with indicator, Ø 2-5 mm, Fluka™
Catalog # 28211-1KG
CAS Number:  
Molecular sieve dehydrate, beads for drying gases, with indicator, Ø 2-5 mm, Fluka™
Catalog # 28211-1KG
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    1 kg
  • Seq:
    4024

Specifications

Specification Test Results

  • Appearance (color):
    complying
  • Loss on ignition (900°C):
    Max. 4.0 %
  • Water adsorption capacity:
    Min. 15 %

Specification Test Results

About this item

 :
Melting Pt:400 °C
Density:2 g/cm³
Storage Temperature:Ambient
MDL Number:MFCD00147630