kr 0,00
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Norway
ContactUsLinkComponent
Rhod-2 AM (Bromide)
Rhod-2 AM (Bromide) fluorescent indicator, for calcium determination
Rhod-2 AM (Bromide) fluorescent indicator, for calcium determination
Rhod-2 AM (Bromide) fluorescent indicator, for calcium determination
Supplier:
Biotium
Certificates
Expand image
Rhod-2 AM (Bromide) fluorescent indicator, for calcium determination
Supplier:
Biotium
About this item
CAS Number:
145037-81-6
:
Formula:
C₅₂H₅₉BrN₄O₁₉
:
1124.0 g/mol
Storage Temperature:
Freezer