Order Entry
Norway
ContactUsLinkComponent
 
BAPTA tetrapotassium salt ≥95% (by TLC) ion chelator, for calcium determination
Catalog # 50002.
Supplier:  Biotium
CAS Number:  
BAPTA tetrapotassium salt ≥95% (by TLC) ion chelator, for calcium determination
Catalog # 50002.
Supplier:  Biotium
CAS Number:  

Specifications

  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :628.79 g/mol
Storage Temperature:Refrigerator
CAS Number:73630-08-7
Formula:C₂₂H₂₀N₂O₁₀K₄
MDL Number:MFCD00070080