Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
kr 0,00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Norway
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Standards
Environmental Analysis Standards
Minerals, (CRM), Supelco®
Minerals, (CRM), Supelco®
Minerals, (CRM), Supelco®
Catalog # QC1041-1KT
Supplier:
Merck
CAS Number:
Utvid bildet
Minerals, (CRM), Supelco®
Catalog # QC1041-1KT
Supplier:
Merck
CAS Number:
Specifications
Pk:
1 KIT
Description:
Minerals
Type:
Certified reference material
Specifications
About this item
Produktinformasjon
Produktinformasjon