Supelco® Organic Reference Standard, n-Nitrosomorpholine solution
Catalog # CRM40485
Supplier: Merck
CAS Number:
Specifications
- Concentration:5000 µg/ml in methanol
- Pk:1 mL
- Standards:Certified reference material
- Description:n-Nitrosomorpholine solution
- Storage conditions:2…8 °C
- Hazard class description:H225 - H301 + H311 + H331 - H351 - H370
- Shipping conditions:WICE
- M-Clarity quality level:MQ100
- Flash point:9,7 °C - Closed cup
- Pk Info:Ampule of 1 ml
Specifications
About this item
- GHS signal: GHS02, GHS06, GHS08
Caution: P201 - P210 - P280 - P301 + P310 + P330 - P302 + P352 + P312 - P304 + P340 + P311