Order Entry
Norway
ContactUsLinkComponent
Supelco® Drug and metabolite standards, methandienone solution, (CRM)
Supelco® Drug and metabolite standards, methandienone solution, (CRM)
Catalog # M-912-1ML
Supplier:  Merck
CAS Number:  
undefined
Supelco® Drug and metabolite standards, methandienone solution, (CRM)
Catalog # M-912-1ML
Supplier:  Merck
CAS Number:  

Specifications

  • Concentration:
    1,0 mg/ml in 1,2-dimethoxyethane
  • Pk:
    1 mL
  • Standards:
    Certified reference material
  • CAS No.:
    72-63-9
  • Description:
    Methandienone solution
  • Description:
    Methandienone solution
  • Pk Info:
    Ampoule of 1 ml

Specifications

About this item