Order Entry
Norway
ContactUsLinkComponent
Supelco® Drug and metabolite standards, O-desmethyl-cis-tramadol hydrochloride solution, (CRM)
Supelco® Drug and metabolite standards, O-desmethyl-cis-tramadol hydrochloride solution, (CRM)
Catalog # T-035-1ML
Supplier:  Merck
CAS Number:  
undefined
Supelco® Drug and metabolite standards, O-desmethyl-cis-tramadol hydrochloride solution, (CRM)
Catalog # T-035-1ML
Supplier:  Merck
CAS Number:  

Specifications

  • Concentration:
    1,0 mg/ml in methanol (as free base)
  • Pk:
    1 mL
  • Standards:
    Certified reference material
  • CAS No.:
    148262-77-5
  • Description:
    O-Desmethyl-cis-tramadol hydrochloride solution
  • Description:
    O-Desmethyl-cis-tramadol hydrochloride solution
  • Pk Info:
    ampoule of 1 ml

Specifications

About this item