Heating tapes, IT-H
Supplier: ISOPAD
Certificates
About this item
Isopad IT-H heating tape is suitable where high power input is needed or where work has to be carried out at high temperatures.
- Working element temperature of up to 900 °C
- Non-hazardous, ordinary area
- Storage temperature –40 to +50 °C
This tape should only be used in dry atmospheres with additional electrical protection and always with a temperature regulator.