Order Entry
Norway
ContactUsLinkComponent
Swabs, PurSwab®
Swabs, PurSwab®
Catalog # PMPC870-PCDBL
Supplier:  PURITAN MEDICAL
undefined
Swabs, PurSwab®
Catalog # PMPC870-PCDBL
Supplier:  PURITAN MEDICAL

Specifications

  • Ø:
    2,2 (Tip); 1,52 (Handle) mm
  • Sterile:
    -
  • Handle length:
    75,5 mm
  • Length:
    76,2 mm
  • Shaft/head:
    Paper/cotton
  • Pk:
    2.500

Specifications

About this item

These double-ended swabs with cotton tips are ideal for cleaning sensitive components in critical environments.

  • Low in NVR
  • Highly absorbent and does not swell in solvents
  • Lint- and residue-free
  • Sharp, double pointed tip with fine paper handle
  • Anti-static packaging contains no silicone
Caution: *The product(s) on this page are outside the scope of the EU Directive 2019/904