R-sulphoraphane ≥98%
Supplier: Cayman Chemical
Certificates
About this item
CAS Number:142825-10-3
:
Formula:C₆H₁₁NOS₂
MDL Number:MFCD09039283
:177.2876 g/mol
Storage Temperature:Freezer
UN:1170
ADR:3,II
:GHS02
Specification Test Results
- Purity:≥98%
- λmax:246 nm