Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
kr 0,00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Norway
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
7-Dehydrocholesterol
7-Dehydrocholesterol ≥90%, crystalline solid
7-Dehydrocholesterol ≥90%, crystalline solid
:
Cayman Chemical
7-Dehydrocholesterol ≥90%, crystalline solid
:
Cayman Chemical
:
434-16-2
:
:
C₂₇H₄₄O
:
MFCD00003624
:
384.64 g/mol
Purity:
≥90%