Order Entry
Norway
Orders LinkContactUsLinkComponent
Oxy setup blend (29 components)
Oxy setup blend (29 components)
Oxy set-up blend, 2 ml prescored ampoule, 1
Catalog # REST33034
Supplier:   Restek
Oxy setup blend (29 components)
Catalog # REST33034
Supplier:  Restek
CAS Number:  
Oxy setup blend (29 components)
Oxy setup blend (29 components)
Oxy set-up blend, 2 ml prescored ampoule, 1
CAS Number:  
Price
kr 5 675,00
Log in for availability
1 * 1 Items
Product Details & Documents
ASTM Method D6730-01 is specifically designed for the determination of the individual hydrocarbons present in spark ignition fuels, as well as fuel blends containing oxygenates such as methyl tert-butyl ether, ethyl tert-butyl ether, tert-butanol, ethanol, etc.
It consists of 29 components, they are benzene (1,00%), tert-butanol (0,50%), cyclohexane (28,9%), n-decane (1,00%), 2,3-dimethylbutane (0,50%), trans-1,2-dimethylcyclopentane (0,50%), 2,3-dimethylheptane, (0,20%), dodecane (0,25%), ethanol (8,00%), ethylbenzene (25,0%) ,3-ethylpentane (0,20%), n-heptane (2,00%), n-hexane (2,00%) ,2-methylbutene-2 (2,50%), methyl tert-butyl ether (10,0%), 1-methylcyclopentene (0,50%), 1-methyl-2-ethylbenzene (0,50%), 1-methylnaphthalene (0,25%), 5-methylnonane, (0,20%), naphthalene (0,50%), n-nonane (2,00%), n-octane (2,00%), n-pentane (2,00%), 1,2,3,5-tetramethylbenzene (0,25%), toluene (7,00%), tridecane (0,25%), 2,3,3-trimethylpentane (0,50%), undecane (0,50%), p-xylene (1,00%),
Specifications
Specifications
  • Pk:
    1
  • Description:
    Oxy set-up blend
  • Pk Info:
    2 ml prescored ampoule