Order Entry
Norway
ContactUsLinkComponent
 
Methylene chloride (dichloromethane)
Catalog # REST30401
Supplier:  Restek
CAS Number:  
undefined
Methylene chloride (dichloromethane)
Catalog # REST30401
Supplier:  Restek
CAS Number:  

Specifications

  • Concentration:
    2000 μg/ml in P and T methanol
  • Pk:
    1 mL
  • CAS No.:
    75-09-2
  • Description:
    Methylene chloride (dichloromethane)
  • Formula:
    CH₂Cl₂
  • Pk Info:
    1 ml/ampoule

Specifications

About this item

Certifications: Certified reference materials (CRMs) manufactured and QC-tested in ISO-accredited labs satisfy your ISO requirements.