Order Entry
Norway
ContactUsLinkComponent
Eriochrome blue black B indicator for complexometry (C.I. 14640), Fluka™
Eriochrome blue black B indicator for complexometry (C.I. 14640), Fluka™
  45540-25G
 :  
Eriochrome blue black B indicator for complexometry (C.I. 14640), Fluka™
  45540-25G
 :  

 

  • Pack type:
    Glass bottle
  • Pk:
    25 g
  • Seq:
    3308

 

 

  • Appearance of the substance:
    complying
  • Absorption maximum:
    523 - 533 nm
  • Absorbtion (1%/1cm):
    Min. 100
  • Suitability for Complexometry:
    complying

 

 

 :
 :416.39 g/mol
 :222-639-6
 :GHS07
 :Ambient
 :3564-14-5
 :C₂₀H₁₃N₂NaO₅S
 :MFCD00003934