Sodium acetate ≥99.0% ACS, Fluka™
Supplier: Honeywell Chemicals
Certificates
About this item
CAS Number:127-09-3
:
Formula:H₃CCOONa
MDL Number:MFCD00012459
:82.03 g/mol
Merck Index:12,08711
Boiling Pt: Dec. (on heating)
Melting Pt:324 °C
Density:1.45 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:204-823-8
REACH:01-2119485123-42
Specification Test Results
- Assay (HClO₄-titr.):Min. 99.0 %
- Water insoluble matter:Max. 0.01 %
- Loss on drying (120°C):Max. 1.0 %
- pH (5 %, 25 °C):7.0 - 9.2
- Calcium (Ca):Max. 0.005 %
- Iron (Fe):Max. 0.001 %
- Magnesium (Mg):Max. 0.002 %
- Heavy metals (as Pb):Max. 0.001 %
- Chloride (Cl):Max. 0.002 %
- Phosphate (PO₄):Max. 0.001 %
- Sulfate (SO₄):Max. 0.003 %
- IR spectrum:complying
- Form:Solid
- pKa :4.76 (acetic acid)