Order Entry
Norway
ContactUsLinkComponent
Calcium hydroxide ≥96.0% (by titrimetric analysis), Purum p.a.
Calcium hydroxide ≥96.0% (by titrimetric analysis), Purum p.a.
Certificates
Calcium hydroxide ≥96.0% (by titrimetric analysis), Purum p.a.

Specification Test Results

  • Assay:
    96.0 - 102.0 %
  • Carbonate (as CaCO₃):
    Max. 4 %
  • Cadmium (Cd):
    Max. 50 ppm
  • Cobalt (Co):
    Max. 50 ppm
  • Copper (Cu):
    Max. 50 ppm
  • Iron (Fe):
    Max. 500 ppm
  • Potassium (K):
    Max. 200 ppm
  • Sodium (Na):
    Max. 100 ppm
  • Nickel (Ni):
    Max. 50 ppm
  • Lead (Pb):
    Max. 50 ppm
  • Zinc (Zn):
    Max. 50 ppm
  • Total S (as SO₄):
    Max. 500 ppm
  • Chloride (Cl):
    Max. 200 ppm

Specification Test Results

Related Information