Calcium hydroxide ≥96%, Puriss. p.a. Reag. Ph. Eur., Fluka™
Supplier: Honeywell Chemicals
Certificates
About this item
CAS Number:1305-62-0
:
Formula:Ca(OH)₂
MDL Number:MFCD00010901
:74.09 g/mol
Merck Index:13,01675
Boiling Pt: Dec. (on heating)
Melting Pt:580 °C
Density:2.24 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:215-137-3
ADR:8,III
:GHS07
:GHS05
Specification Test Results
- Assay:Min. 96 %
- Assay of CaCO₃:Max. 3 %
- Insoluble in hydrochloric acid:Max. 0.03 %
- Copper (Cu):Max. 0.0005 %
- Iron (Fe):Max. 0.05 %
- Lead (Pb):Max. 0.0002 %
- Zinc (Zn):Max. 0.0005 %
- Substances not precipitated by NH₄OH (asO₄):Max. 2.5 %
- Chloride (Cl):Max. 0.005 %
- Sulfate (SO₄):Max. 0.05 %