Amidosulphonic acid ≥99.5% (by alkalimetry)
Supplier: Honeywell Chemicals
Certificates
About this item
CAS Number:5329-14-6
:
Formula:H₃NO₃S
MDL Number:MFCD00011603
:97.09 g/mol
Merck Index:13,09006
Melting Pt:209 °C.205 °C
Density:2.1 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:226-218-8
UN:2967
ADR:8,III
:GHS07
Specification Test Results
- Assay (alkalimetric):Min. 99.5 %
- Residue on ignition:Max. 0.02 %
- Iron (Fe):Max. 0.003 %
- Heavy metals (as Pb):Max. 0.001 %
- Chloride (Cl):Max. 0.001 %
- Sulfate (SO₄):Max. 0.15 %