Ammonium chloride ≥99.5%, Puriss. p.a. ACS, ISO, Reag. Ph. Eur., Fluka™
Supplier: Honeywell Chemicals
Certificates
About this item
CAS Number:12125-02-9
:
Formula:NH₄Cl
MDL Number:MFCD00011420
:53.49 g/mol
Merck Index:13,00510
Melting Pt:338 °C (dec.)
Density:1.536 g/cm³ (20 °C)
EINECS:235-186-4
:GHS07
Storage Temperature:Ambient
Specification Test Results
- Assay:Min. 99.5 %
- Assay (calculated to the dried substance):99.5 - 100.5 %
- Water insoluble matter:Max. 0.005 %
- Loss on drying (105°C):Max. 0.5 %
- Residue on ignition (SO₄):Max. 0.01 %
- pH (5 %, 25 °C):4.5 - 5.5
- Acidity or alkalinity:complying
- Arsenic (As):Max. 0.5 ppm
- Calcium (Ca):Max. 5 ppm
- Copper (Cu):Max. 2 ppm
- Iron (Fe):Max. 2 ppm
- Potassium (K):Max. 0.005 %
- Magnesium (Mg):Max. 5 ppm
- Vapor Pressure:1 mmHg ( 160.4 °C)
- Sodium (Na):Max. 0.005 %
- Nickel (Ni):Max. 0.0001 %
- Lead (Pb):Max. 1 ppm
- Zinc (Zn):Max. 0.0002 %
- Heavy metals (ICP) ACS:Max. 5 ppm
- Nitrate (NO₃):Max. 0.0005 %
- Phosphate (PO₄):Max. 2 ppm
- Sulfate (SO₄):Max. 20 ppm
- Iodide and bromide:complying
- Appearance of the solution:complying