Order Entry
Norway
ContactUsLinkComponent
 
Tin(II) chloride dihydrate ≥98%, Puriss. p.a. ACS, ISO, Reag. Ph. Eur., Fluka™
 
Tin(II) chloride dihydrate ≥98%, Puriss. p.a. ACS, ISO, Reag. Ph. Eur., Fluka™
 

 

  • Assay:
    98.0 - 103.0 %
  • Solubility in HCl:
    complying
  • Ammonium (NH₄):
    Max. 0.002 %
  • Arsenic (As):
    Max. 0.0001 %
  • Calcium (Ca):
    Max. 0.005 %
  • Copper (Cu):
    Max. 0.001 %
  • Iron (Fe):
    Max. 0.002 %
  • Potassium (K):
    Max. 0.005 %
  • Magnesium (Mg):
    Max. 0.005 %
  • Manganese (Mn):
    Max. 0.0005 %
  • Sodium (Na):
    Max. 0.005 %
  • Nickel (Ni):
    Max. 0.0005 %
  • Lead (Pb):
    Max. 0.005 %
  • Other heavy metals (as Pb):
    Max. 0.01 %
  • Sulfate (SO₄):
    Max. 0.002 %
  • Substances not precipitated by H₂S (SO₄):
    Max. 0.05 %